Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ DNMT3A Assay Kit

Catalog No. 89027040 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-027-040 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-027-040 Supplier Abnova™ Supplier No. KA0683
Add to Cart
Add to Cart

For measuring the amount of DNMT3A

Specifications

Content And Storage Store DA3 and DA5 at -20°C away from light. Store DA1, DA2, DA4, DA6, Blocking buffer and 8 well strips at 4°C away from light. Store other components at room temperature.
Description DNMT3A Assay Kit is use for measuring the amount of DNMT3A.
Detection Method Colorimetric
For Use With (Application) Quantification
Includes 25mL Wash Buffer (10X) (DA1), 3mL Assay Buffer (DA2), 20μL Dnmt3A standard (DA3), 50μL affinity Antibody (DA4), 20μL detection Antibody (DA5), 12mL developing solution (DA6), 6mL stop solution (DA7), 1
Quantity 1 Kit
Type Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.