Learn More
Specifications
Specifications
| Content And Storage | Store the kit at -80°C temporarily. For best results, store the components as described in the protocol. |
| Description | DPP (IV) Inhibitor Screening Assay Kit is a fluorescence-based method for screening DPP (IV) inhibitors. |
| Detection Method | Fluorometric |
| For Use With (Application) | Functional Study |
| Includes | 1 vial DPP assay buffer 10X, 2 vials DPP IV (human recombinant), 1 vial DPP substrate, 1 plate half volume 96-well plate (white), 1 cover 96-well cover sheets |
| Quantity | 1 Kit |
| Type | Inhibitor Screening Assay Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.