Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Esm1 (Mouse) ELISA Kit

Catalog No. 89139258 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
89-139-258 1 Kit
1 options

Catalog No. 89-139-258

Supplier: Abnova™ KA4393

Only null left
Add to Cart
Add to Cart

Sandwich enzyme immunoassay for the quantitative measurement of mouse Esm1 in cell culture supernates, plasma (heparin, EDTA) and serum samples

  • Sample Type: Cell culture supernates, plasma (heparin, EDTA) and serum
  • Sample Volume: 100μL
  • Conjugate: Biotin
  • Reactivity: Mouse
  • Detection Method: Colorimetric
  • Calibration Range: 23.4 to 1500 pg/mL
  • Limit of Detection: <10 pg/mL

  • Applications: Quantification
  • Storage: Store the kit at -20°C.

Specifications

Assay Sensitivity <10 pg/mL
Calibration Range 23.4 to 1500 pg/mL
Description Esm1 (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Esm1.
Detectable Analytes Esm1
Assay Sandwich ELISA
Conjugate Biotin
Cross Reactivity No detectable cross-reactivity with other relevant proteins
Detection Method Colorimetric
Immunoassay Kit Format 96-well Plate
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
Specificity Natural and recombinant mouse ESM1
Sufficient For 96 Assays
For Use With (Application) Quantification
Gene ID (Entrez) 71690
Host Species Mouse
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Volume 100 μL
Storage Requirements Store the kit at -20°C
Show More Show Less

For research use only (RUO).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.