Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Estradiol ELISA Kit

Catalog No. 89100172 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CCS, Saliva
Dried Fecal Extract, Tissue Culture Medium, Urine
Plasma, Serum
Saliva
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-172 CCS, Saliva
89-028-719 Saliva
89-100-717 Plasma, Serum
89-100-931 Dried Fecal Extract, Tissue Culture Medium, Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 89-100-172 Supplier Abnova™ Supplier No. KA0297
Add to Cart
Add to Cart

Includes

  • Wash Concentrate concentrate for 1200mL, 1 x 30mL
  • Substrate ready to use, Tetramethylbenzidine (TMB), 1 x 25mL
  • Stop Solution ready to use, containing 0.5M H2SO4, 1 x 14mL
  • Standard A, ready to use, 0pg/mL,1 x 1mL
  • Standard B ready to use, 1pg/mL, 1 x 1mL
  • Standard C ready to use, 5pg/mL, 1 x 1mL
  • Standard D ready to use, 10pg/mL,1 x 1mL
  • Standard E ready to use, 50pg/mL,1 x 1mL
  • Standard F ready to use, 100pg/mL, 1 x 1mL
  • Microtiter wells 12 x 8 (break apart) strips, 96 wells; Wells coated with an anti-Estradiol antibody (polyclonal), 1 x 96 wells
  • Enzyme Conjugate ready to use, 1 x 26mL
  • Control 1 ready to use, 1 x 1mL
  • Control 2 ready to use, 1 x 1mL

Enzyme Immunoassay for the quantitative measurement of active free Estradiol in Saliva

Specifications

Calibration Range 29.3 to 30000pg/mL
Description 17 beta-Estradiol ELISA Kit is a competitive immunoassay for the quantitative determination of 17 beta-Estradiol in biological and environmental samples.
Detection Method Colorimetric
Quantity 1 Kit
Sample Type CCS, Saliva
Sample Volume 100 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Estradiol ELISA kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.