Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Estrone ELISA Kit

Catalog No. 89028720 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Dried Fecal Extract, Tissue Culture Medium, Urine
Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-028-720 Serum
89-100-932 Dried Fecal Extract, Tissue Culture Medium, Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-028-720 Supplier Abnova™ Supplier No. KA1908
Add to Cart
Add to Cart

Enzyme Immunoassay for the quantitative measurement of Estrone in Human Serum

Specifications

Conjugate HRP
Cross Reactivity Human
Description Estrone ELISA Kit is an enzyme immunoassay for the quantitative measurement of estrone in human serum.
Detection Limit 10.0pg/mL
Detection Method Colorimetric
Includes 30mL Wash concentrate, 25mL Substrate, 14mL stop solution, 1mL standard A, 1mL standard A, 1mL standard A, 1mL standard A, 1mL standard A, 1mL standard A, 96-microtiterwell, 26mL Enzyme Conjugate, 1mL control 1, 1mL control 2
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Serum
Sample Volume 50 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Estrone ELISA kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.