Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Estrone ELISA Kit

Catalog No. 89100932 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Dried Fecal Extract, Tissue Culture Medium, Urine
Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-932 Dried Fecal Extract, Tissue Culture Medium, Urine
89-028-720 Serum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-932 Supplier Abnova™ Supplier No. KA3395
Add to Cart
Add to Cart

Enzyme Immunoassay for the quantitative measurement of Estrone in Human Serum

Specifications

Calibration Range 0 to 2000pg/mL
Conjugate HRP
Description Estrone ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative detection of estrone dried fecal, urine and for tissue culture samples.
Detection Limit 28.2pg/mL
Detection Method Colorimetric
Includes One Clear, break-apart 1 by 8 strip well plastic microtiter plate(s) coated with goat anti-rabbit IgG, Estrone Standard: Estrone at 20,000 pg/mL in a special stabilizing solution (125μL), Estrone Antibody: A rabbit polyclonal antibody specific for estrone (3mL), Estrone Conjugate: A estrone-peroxidase conjugate in a special stabilizing solution (3mL), Assay Buffer Concentrate: A 5X concentrate that should be diluted with deionized or distilled water (28mL), Wash Buffer Concentrate: A 20X concentrate that should be diluted with deionized or distilled water (30mL), TMB Substrate (11mL), Stop Solution: A 1M solution of hydrochloric acid. CAUSTIC (5mL), One Plate Sealer
Quantity 96 Assays
Regulatory Status For research use only (RUO)
Sample Type Dried Fecal Extract, Tissue Culture Medium, Urine
Sample Volume 50 μL
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type Estrone ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.