Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ F10 (Human) Chromogenic Activity Assay Kit

Catalog No. 89028047 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-047 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-047 Supplier Abnova™ Supplier No. KA0972
Add to Cart
Add to Cart

To determine Human F10 activity in plasma and cell culture, functional study

Specifications

Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
Description F10 (Human) Chromogenic Activity Assay Kit is developed to determine human F10 activity in plasma, serum, urine and cell culture samples.
For Use With (Application) Functional Study
Includes 96-well microplate, 10mL samples diluent 6X, 10mL assay diluent, 1 vial RVV, 10μg human FX standard, 2 vials FXa substrate
Quantity 1 Kit
Type Chromogenic Activity Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.