Learn More
Abnova™ F7 (Human) Chromogenic Activity Assay Kit
Catalog No. 89028048 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
89-028-048 1 Kit
1 options
Catalog No. 89-028-048 Supplier Abnova™ Supplier No. KA0971
Add to Cart
Add to Cart
To determine Human F7 activity in plasma and cell culture supernatants, functional study
Specifications
Content And Storage Store the kit at -20°C. Store the Standard, FX, rhTF and FXa Substrate at -20°C. Store other components at 4°C. Avoid repeated freezing and thawing.
Description F7 (Human) Chromogenic Activity Assay Kit is developed to determine human F7 activity in plasma and cell culture supernatants.
For Use With (Application) Functional Study
Includes 96-well antibody coated microwells, 1.6 IU human FVII standard, 20mL EIA diluent concentrate 6X, 30mL wash buffer concentrate 20X, 20mL assay diluent, 1 vial rhTF (Lipoprotein), 1 vial human FX, 2 vials FXa substrate
Quantity 1 Kit
Type Chromogenic Activity Assay Kit