Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Fibrinogen (Rat) ELISA Kit

Catalog No. 89100219 Shop All Abnova Corporation Products
Change view
Click to view available options
Conjugate:
Biotin
HRP
Sample Volume:
25 μL
50 μL
100 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Conjugate Sample Volume
89-100-219 Biotin 50 μL
89-024-525 Biotin 25 μL
89-100-694 HRP 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 89-100-219 Supplier Abnova™ Supplier No. KA0505
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Rat Fibrinogen in Plasma

Specifications

Calibration Range 3.125 to 200ng/mL
Conjugate Biotin
Cross Reactivity Rat
Description Fibrinogen (Rat) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of rat fibrinogen.
Detection Limit 2ng/mL
Detection Method Colorimetric
Includes 96 wells antibody coated wells, Rat FBG standard (200ng), Biotinylated Rat FBG antibody (1 vial), MIX diluent concentration (30mL), Wash Buffer Concentrate 20X (2 x 30mL ), 80 Streptavidin-Peroxidase Conjugate (SP Conjugate), Chromogen Substrate (8mL), Stop Solution (12mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type CCS, Urine
Sample Volume 50 μL
Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
For Use With (Application) Quantification
Product Type Fibrinogen (Rat) ELISA kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.