Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ FIGF (Human) ELISA Kit

Catalog No. 89100487 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CCS, Plasma, Serum, Tissue Sample, Urine
Cell Lysates, Plasma, Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-487 Cell Lysates, Plasma, Serum
89-100-823 CCS, Plasma, Serum, Tissue Sample, Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-487 Supplier Abnova™ Supplier No. KA2252
Add to Cart
Add to Cart

Includes

  • 96-Well Microplate or Strips Coated w/Capture Antibody
  • Biotin-Conjugated Detection Antibody, Lyophilized (2.4μg), Yellow container
  • Avidin-HRP Conjugate, 10μL, Clear container
  • Cytokine Protein Standard, Lyophilized (6.5ng), Red container
  • Ready-to-Use Substrate, 11mL, Brown container
  • Stop Solution, 11mL, Clear container
  • Adhesive Plate Sealers, 4 Sheets
  • Wash Buffer (10X), 50mL, Clear container
  • Protein Standard Diluent, 15mL, Clear container
  • Detection Antibody Diluent, 15mL, Clear container
  • HRP Diluent, 15mL, Clear container

This particular immunoassay utilizes the quantitative technique of a “Sandwich” Enzyme-Linked Immunosorbent Assay (ELISA) where the target protein (antigen) is bound in a “sandwich” format by the primary capture antibodies coated to each well-bottom and the secondary detection antibodies addedsubsequently by the investigator

Specifications

Conjugate Biotin
Cross Reactivity Human
Description FIGF (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human FIGF.
Detection Method Colorimetric
Includes Antibody coated microwells (96 wells), Biotin-conjugated detection antibody (1 vial), Avidin-HRP conjugate (10μL), Cytokine protein standard (1 vial); Ready-to-use substrate (11mL), Stop solution (11mL), Wash buffer 10X (50mL), Protein standard diluent (15mL), Detection antibody diluent (15mL), HRP diluent (15mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell Lysates, Plasma, Serum
Sample Volume 100 μL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.