Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Galactose Assay Kit

Catalog No. 89028401 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-401 100 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-401 Supplier Abnova™ Supplier No. KA1669
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Assay buffer, 10mL
  • Enzyme mix, 120μL
  • Dye reagent, 120μL
  • Standard, 1mL 10mM galactose

For quantitative colorimetric galactose determination in biological sample, culture medium, milk, plasma, saliva, serum, urine

Specifications

Calibration Range 10 to 1000μM colorimetric assay, 10 to 100μM fluorimetric assay
Content And Storage Store the kit at -20°C.
Description Galactose Assay Kit is a quantitative colorimetric galactose determination.
Detection Method Colorimetric
For Use With (Application) Functional Study
Includes 10mL assay buffer, enzyme mix (Dried) 1 vial, 120μL dye reagent, 1mL standard
Quantity 100 Assays
Sample Type Biological Sample, Culture Medium, Milk, Plasma, Saliva, Serum, Urine
Sample Volume 20 μL
Type Activity Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.