Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ General Gene Knockdown Quantification Kit

Catalog No. 89028236 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-236 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-236 Supplier Abnova™ Supplier No. KA1542
Add to Cart
Add to Cart

General Gene Knockdown Quantification Kit is used for measuring gene knockdown induced by siRNA or antisense oligonucleotide at the protein level.

  • Quick and efficient procedure; completion of entire assay needs only 4 hours
  • Innovative colorimetric assay without the need for radioactivity, electrophoresis, or chromatography
  • Internal control is conveniently included for standardization and normalization of target gene knockdown effect
  • Strip microplate format makes the assay flexible, manual or high throughput
  • Simple, reliable, and consistent assay conditions
  • Kit is shipped in two parts, one at ambient room temperature and the second frozen on ice packs at 4°C

Specifications

Description General Gene Knockdown Quantification Kit is used for measuring gene knockdown induced by siRNA or antisense oligonucleotide at the protein level.
For Use With (Application) Quantification
Sample Volume 10μL
Content And Storage Store the detection antibody at -20°C. Store Q2, Q4, Q6, GAPDH control antibody and 8-well assay strips at 4°C away from light. Store all other components at room temperature.
Product Type General gene knockdown quantification kit
Quantity Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.