Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Global Acetyl Histone H4K16 Quantification Kit (Fluorometric)

Catalog No. 89028229 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-229 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-229 Supplier Abnova™ Supplier No. KA1535
Add to Cart
Add to Cart

To measure global acetylation of histone H4K16

Specifications

Calibration Range 20 to 5000ng/well
Content And Storage Store F4 and the standard at -20°C. Store all other components at 4°C away from light.
Description Global Acetyl Histone H4K16 Quantification Kit (Fluorometric) is used for measuring global acetylation of histone H4K16.
Detection Method Fluorometric
For Use With (Application) Quantification
Includes 10mL F1 (wash buffer 10X), 6mL F2 (antibody buffer), 5μL F3 (detection antibody, 1mg/mL), 12μL F4 (Fluoro developer), 12μL F5 (Fluoro enhancer), 4mL F6 (Fluoro dilution), 10μL standar
Quantity 1 Kit
Sample Type Histone Extract
Sample Volume 50 μL
Type Quantification Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.