Learn More
Abnova™ Global Tri-Methyl Histone H3-K36 Quantification Kit (Colorimetric)
Shop All Abnova Corporation ProductsSpecifications
Specifications
| Description | Global Tri-Methyl Histone H3-K36 Quantification Kit (Colorimetric) is use for measuring global tri-methylation of histone H3-K36. |
| Detection Method | Colorimetric |
| Includes | 20mL C1 (Wash Buffer 10X), 12mL C2 (Antibody Buffer), 10μL C3 (detection Antibody, 1mg/mL), 10μL C4 (color developer), 6mL C5 (stop solution), 20μL standard control (100μg/mL), 98-wel |
| Quantity | 1 Kit |
| Content And Storage | Store C3 and standard control at -20°C. Store all other components at 4°C away from light. |
| For Use With (Application) | Quantification |
| Product Type | Global Tri Methyl Histone H3-K36 quantification kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.