Learn More
Description
Includes
- Reagent, 50mL
- Standard, 1mL (300mg/dL)
Specifications
Specifications
| Calibration Range | 0.7 to 300mg/dL |
| Content And Storage | Store the reagent at room temperature and standard at -20°C. |
| Description | Glucose Assay Kit is a quantitative colorimetric glucose determination at 630 nm. |
| Detection Method | Colorimetric |
| For Use With (Application) | Functional Study |
| Includes | 50mL Reagent, 1mL 300mg/dL standard |
| Quantity | 1 Kit |
| Sample Type | Biological Sample, Plasma, Serum |
| Sample Volume | 5 μL |
| Type | Activity Detection Assay Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.