Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Glutamate Assay Kit

Catalog No. 89028402 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-402 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-402 Supplier Abnova™ Supplier No. KA1670
Add to Cart
Add to Cart

Includes

  • Assay buffer, 10mL
  • NAD solution, 1mL
  • Enzyme mix, 120μL
  • MTT solution, 1.5mL
  • Standard, 1mL (100mM glutamate)

For quantitative colorimetric determination of glutamate at 565nm in food sample, plasma, serum, tissue

Specifications

Calibration Range 50 to 2500μM
Content And Storage Store the kit at -20°C.
Description Glutamate Assay Kit is a quantitative colorimetric determination of glutamate at 565 nm.
Detection Method Colorimetric
For Use With (Application) Functional Study
Includes 10mL assay buffer, 1mL NAD solution, 120μL enzyme mix, 1.5mL MTT Solution, 1mL 100mM Glutamate standard
Quantity 1 Kit
Sample Type Food Sample, Plasma, Serum, Tissue
Sample Volume 20 μL
Type Activity Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.