Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Glutamate Dehydrogenase Assay Kit

Catalog No. 89101203 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Culture Medium, Plasma, Serum, Tissue, Urine
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-101-203 Culture Medium, Plasma, Serum, Tissue, Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-101-203 Supplier Abnova™ Supplier No. KA3787
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • GDH assay buffer, 25mL
  • Glutamate (2M), 1mL
  • GDH developer (lyophilized), 1 vial
  • GDH positive control (lyophilized), 1 vial
  • NADH (0.5μmol, lyophilized), 1 vial

Used for detecting GDH activity and functional study

Specifications

Calibration Range 0.4 to 80μ/L
Content And Storage Store the kit at -20°C.
Description Glutamate dehydrogenase Assay Kit is a quantitative colorimetric determination of glutamate dehydrogenase at 565 nm.
Detection Method Colorimetric
For Use With (Application) Functional Study
Quantity 1 Kit
Sample Type Culture Medium, Plasma, Serum, Tissue, Urine
Sample Volume 20 μL
Type Activity Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.