Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Glutathione Peroxidase Assay Kit

Catalog No. 89028360 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-360 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-360 Supplier Abnova™ Supplier No. KA1628
Add to Cart
Add to Cart

For quantitative colorimetric glutathione peroxidase determination in biological sample

Specifications

Calibration Range 40 to 800U/L
Content And Storage Store the kit at -20°C.
Description Glutathione Peroxidase Assay Kit is a quantitative colorimetric glutathione peroxidase determination.
Detection Method Colorimetric
For Use With (Application) Functional Study
Includes 25mL assay buffer, 1mL GR enzyme, 240μL Glutathione, 40μL NADPH, 100μL calibrator, 50μL Cumene Hydroperoxide, 9μL positive control Glutathione Peroxidase (GPX)
Quantity 1 Kit
Sample Type Biological Sample
Sample Volume 10 μL
Type Activity Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.