Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Glutathione S-Transferase Assay Kit

Catalog No. 89024602 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-602 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-602 Supplier Abnova™ Supplier No. KA1316
Add to Cart
Add to Cart

Used for assaying the activity of GST and functional study

Specifications

Content And Storage Store the kit at -20°C.
Description Glutathione S-Transferase Assay Kit is a colorimetric method for assaying the activity of GST.
Detection Method Colorimetric
For Use With (Application) Functional Study
Includes 1 vial GST assay buffer 2X, 1 vial GST sample buffer 2X, 1 vial GST assay (control), 1 vial GST Glutathione, 1 vial GST CDNB, 1 plate 96-well plate (colorimetric assay), 1 cover 96-well cover sheet
Quantity 1 Kit
Type Activity Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.