Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Glycerol Cell-Based Assay Kit

Catalog No. 89024603 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-603 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-603 Supplier Abnova™ Supplier No. KA1317
Add to Cart
Add to Cart

Includes

  • Glycerol standard solution, 1 vial
  • Free glycerol assay reagent (10x), 2 vials
  • Chloroquine positive control (25mM), 1 vial

Used for quantifying glycerol and functional study

Specifications

Content And Storage Store the kit at -20°C temporarily. For best results, store the components as described in the protocol.
Description Glycerol Cell-Based Assay Kit is a colorimetric method for quantifying glycerol.
Detection Method Colorimetric
For Use With (Application) Functional Study
Includes 1 vial glycerol standard solution, 2 vials free glycerol assay reagent 10X, 1 vial chloroquine positive control (25mM)
Quantity 1 Kit
Type Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.