Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ GRN (Human) ELISA Kit

Catalog No. 89100484 Shop All Abnova Corporation Products
Change view
Click to view available options
Conjugate:
Biotin
Sample Type:
Cell Lysates, Plasma, Serum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Conjugate Sample Type
89-100-484 Biotin Cell Lysates, Plasma, Serum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-484 Supplier Abnova™ Supplier No. KA2249
Add to Cart
Add to Cart

Includes

  • Microstrips Coated w/Capture Antibody, 12 x 8 well Microstrips
  • Protein Standard, Lyophilized (100ng), Red
  • Biotinylated Detection Antibody, Lyophilized (1 vial), Yellow
  • 400x Streptavidin-HRP, 30μL, Blue
  • Wash Buffer (10x), 50mL, Clear
  • Assay Diluent, 50mL, Clear
  • Ready-to-Use Substrate, 12mL, Brown
  • Stop Solution, 12mL, Clear
  • Adhesive Plate Sealers, 4 Sheets

Use to measure quantitatively human Progranulin. Sandwich enzyme immunoassay.

This particular immunoassay utilizes the quantitative technique of a “Sandwich” Enzyme-Linked Immunosorbent Assay (ELISA) where the target protein (antigen) is bound in a “sandwich” format by the primary capture antibodies coated to each well-bottom and the secondary detection antibodiesadded subsequently by the investigator

Specifications

Assay Sensitivity 63 to 4000 pg/mL
Calibration Range 62.5 to 4000pg/mL
Content And Storage Store the kit at 4°C
Description Progranulin (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human Progranulin.
Conjugate Biotin
Cross Reactivity Human, Mouse
Detection Method Colorimetric
Product Type ELISA Kit
Sample Type Cell Lysates, Plasma, Serum
Sufficient For 96
For Use With (Application) Quantification
Kit Contents Antibody coated microwells (96 wells), Biotin-conjugated detection antibody (1 vial), Avidin-HRP conjugate (10μL), Cytokine protein standard (1 vial); Ready-to-use substrate (11mL), Stop solution (11mL), Wash buffer 10X (50mL), Protein standard diluent (15mL), Detection antibody diluent (15mL), HRP diluent (15mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Volume 100 μL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.