Learn More
Description
Includes
- HAMA Microtiter plate Pre-coated with mouse IgG, 96-wells
- HRP Conjugate HRP conjugate to Mouse IgG, 12mL
- HAMA Standard: Rabbit anti Mouse IgG antibody (800ng/vial) in a buffered protein base with Preservative, lyophilized, 2 vials
- Calibrator diluent Animal serum with buffer and preservative, 25mL
- Wash Buffer (20x) 20-fold concentrated solution of buffered surfactant, 60mL
- Substrate A: Buffered solution with H2O2, 10mL
- Substrate B: Buffered solution with TMB, 10mL
- Stop solution 2N Sulphuric Acid (H2SO4) (Caution: Caustic Material), 15mL
Specifications
Specifications
| Calibration Range | 6.25 to 400 ng/mL |
| Content And Storage | Store the kit at 4°C |
| Description | HAMA (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human anti-mouse antibody. |
| Conjugate | HRP |
| Cross Reactivity | Human |
| Detection Method | Colorimetric |
| Product Type | HAMA (Human) ELISA kit |
| Sample Type | Serum |
| For Use With (Application) | Quantification |
| Kit Contents | 96-well HAMA microtiter plate, 12mL HRP Conjugate, 2 vial HAMA standard, 25mL calibrator Diluent, 60mL Wash Buffer (20X), 10mL Substrate A, 10mL Substrate B, 15mL stop solution |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.