Learn More
Specifications
Specifications
| Content And Storage | Store the kit at -80°C temporarily. For best results, store the components as described in the protocol. |
| Description | HDAC Cell-Based Assay Kit is a fluorescence-based method for assaying the activity of HDAC . |
| Detection Method | Fluorometric |
| For Use With (Application) | Functional Study |
| Includes | 5mL HDAC assay buffer 10X, 500μL Nonidet P-40 assay reagent (10%), 400μL HDAC Deacetylated standard, 50μL HDAC1 positive control, 100μL HDAC substrate, 20mg HDAC developer, 250 |
| Quantity | 1 Kit |
| Type | Cell-Based Assay Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.