Learn More
Abnova™ HDAC2 Assay Kit
Catalog No. 89027045 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
89-027-045 1 Kit
1 options
Catalog No. 89-027-045 Supplier Abnova™ Supplier No. KA0688
Add to Cart
Add to Cart
Includes
  • HB1 (10X wash buffer), 22mL
  • HB2 (HDAC assay buffer), 2mL
  • HB3 (Blocking buffer), 20mL
  • HB4 (Capture antibody, 200μg/mL), 26μL
  • HB5 (Detection antibody 200μg/mL), 20μL
  • HB6 (Developing solution), 12mL
  • HB7 (Stop solution), 6mL
  • HDAC2 control (100ng/μL), 32μL
  • 8-Well assay strip (with Frame), 12
Used for measuring the amount of HDAC2
Specifications
Content And Storage Store HB5 and HDAC2 control at -20°C. Store HB3, HB4, HB6 and 8 well strips at 4°C away from light. Store all other components at room temperature.
Description HDAC2 Assay Kit is use for measuring the amount of HDAC2.
For Use With (Application) Quantification
Includes 22mL HB1 (Wash Buffer 10X), 2mL HB2 (HDAC Assay Buffer), 20μL HB3 (blocking Buffer), 26μL HB4 (capture Antibody, 200μg/mL), 20μL HB5 (detection Antibody 200μg/mL), 1
Quantity 1 Kit
Type Detection Assay Kit