Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ HDAC8 Assay Kit

Catalog No. 89027191 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-027-191 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-027-191 Supplier Abnova™ Supplier No. KA0631
Add to Cart
Add to Cart

Includes

  • HC1 (10X wash buffer), 22mL
  • HC2 (HDAC assay buffer), 2mL
  • HC3 (blocking buffer), 20mL
  • HC4 (capture antibody 200μg/mL), 26μL
  • HC5 (detection antibody 200μg/mL), 20μL
  • HC6 (developing solution), 12mL
  • HC7 (stop solution), 6mL
  • HDAC8 control (100ng/μL), 24μL
  • 8 well assay strip (with frame), 12

Use for measuring the amount of HDAC1

Specifications

Content And Storage Store HC5 and HDAC8 control at -20°C. Store HC3, HC4, HC6 and 8 well strips at 4°C away from light. Store all other components at room temperature.
Description HDAC8 Assay Kit is use for measuring the amount of HDAC1.
Detection Method Colorimetric
For Use With (Application) Quantification
Includes 22mL HC1 (Wash Buffer 10X), 2mL HC2 (HDAC Assay Buffer), 20mL HC3 (blocking Buffer), 26μL HC4 (capture Antibody 200μg/mL), 20μL HC5 (detection Antibody 200μg/mL), 12mL HC6 (developing
Quantity 1 Kit
Type Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.