Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ HDL and LDL/VLDL Assay Kit

Catalog No. 89028403 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-403 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-403 Supplier Abnova™ Supplier No. KA1671
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Quantitative colorimetric determination of HDL and LDL/VLDL cholesterol

  • Sample Volume: Serum
  • Sample Volume: 20μL

Functional Study

Specifications

Calibration Range 5 to 300mg/dL
Content And Storage Store the kit at -20°C. Avoid repeated freezing and thawing.
Description HDL and LDL/VLDL Assay Kit is a quantitative colorimetric determination of HDL and LDL/VLDL cholesterol.
Detection Method Colorimetric
For Use With (Application) Functional Study
Includes 1.5mL PBS, 1.5mL Precipitation reagent, 20mL assay buffer, 120μL enzyme mix, 2mL NAD Solution, 1mL standard 300mg/dL Cholesterol
Quantity 1 Kit
Sample Type Serum
Sample Volume 20 μL
Type Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.