Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Heme Assay Kit

Catalog No. 89028349 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-349 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-349 Supplier Abnova™ Supplier No. KA1617
Add to Cart
Add to Cart

Includes

  • Reagent, 50L
  • Calibrator: (equivalent to 62.5μM heme), 10mL

Colorimetric determination of total heme at 400nm in blood, plasma, serum, urine

Specifications

Calibration Range 0.6 to 125μM
Content And Storage Store the kit at 4°C.
Description Heme Assay Kit is a colorimetric determination of total heme at 400 nm.
Detection Method Colorimetric
For Use With (Application) Functional Study
Includes 50mL Reagent, 10mL calibrator (equivalent to 62.5μM Heme)
Quantity 1 Kit
Sample Type Blood, Plasma, Serum, Urine
Sample Volume 100 μL
Type Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.