Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Histone Demethylase LSD1 Inhibitor Screening Assay Core Kit

Catalog No. 89028242 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-242 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-242 Supplier Abnova™ Supplier No. KA1548
Add to Cart
Add to Cart

Includes

  • HH1 (LSD1 assay Bbffer), 4mL
  • HH2 (LSD1 substrate, 0.7mM), 300μL
  • HH3 (LSD1 assay standard, 100mM), 20μL
  • HH4 (LSD1 inhibitor, 1mM), 50μL
  • HH5 (Fluoro developer), 24μL
  • HH6 (Fluoro enhancer), 24μL
  • HH7 (Fluoro diluter), 8mL
  • Twelve 8 well standard control strips

Used for screening histone demethylase LSD1 inhibitors

Specifications

Content And Storage Store HH2 and HH5 at -20°C away from light. Store all other components at 4°C away from light.
Description Histone Demethylase LSD1 Inhibitor Screening Assay Core Kit is use for screening histone demethylase LSD1 inhibitors.
Detection Method Fluorometric
For Use With (Application) Functional Study
Includes 4mL HH1 (LSD1 assay buffer), 300μL HH2 (LSD1 substrate, 0.7mM), 20μL HH3 (LSD1 assay standard, 100mM), 50μL HH4 (LSD1 inhibitor, 1mM), 24μL HH5 (Fluoro developer), 24μ
Quantity 1 Kit
Sample Type Purified Enzyme (Active)
Type Inhibitor Screening Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.