Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Hydrogen Peroxide Cell-Based Assay Kit

Catalog No. 89024608 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-608 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-608 Supplier Abnova™ Supplier No. KA1322
Add to Cart
Add to Cart

Used for quantifying hydrogen peroxide and functional study

Specifications

Content And Storage Store the kit at -20°C temporarily. For best results, store the components as described in the protocol.
Description Hydrogen Peroxide Cell-Based Assay Kit is a fluorescence-based method for quantifying hydrogen peroxide.
Detection Method Fluorometric
For Use With (Application) Functional Study
Includes 1 vial buffer tablet, 100μL hydrogen peroxide standard, 100μL hydrogen peroxide detector ADHP, 100μL horseradish peroxidase, 2mg catalase
Quantity 1 Kit
Type Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.