Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Hydroxymethylated DNA Quantification Kit (Fluorometric)

Catalog No. 89028208 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-208 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-208 Supplier Abnova™ Supplier No. KA1513
Add to Cart
Add to Cart

To measure hydroxymethylated DNA

Specifications

Calibration Range 20 to 200ng
Content And Storage Store HF3, HF4, HF5, HF7, HF8 and HF9 at -20°C away from light. Store HF1, HF6, HF10 and 8-Well Assay Strips at 4°C away from light. Store HF2 and HF11 at room temperature away from light.
Description Hydroxymethylated DNA Quantification Kit (Fluorometric) is used for measuring hydroxymethylated DNA
Detection Method Fluorometric
For Use With (Application) Quantification
Includes 14mL HF1 (wash buffer 10X), 5mL HF2 (binding solution), 10μL HF3 (negative control I, 20μg/mL), 10μL HF4 (negative control II, 20μg/mL), 10μL HF5 (positive control,
Quantity 1 Kit
Sample Type DNA Extract
Sample Volume 1 to 8 μL
Type Quantification Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.