Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ IFNG (Mouse) ELISA Kit

Catalog No. 89100753 Shop All Abnova Corporation Products
Catalog No. 89-100-753 Supplier Abnova™ Supplier No. KA3046
Add to Cart
Add to Cart

Includes

  • One 96-well plate pre-coated with anti-Mouse IFNγ antibody, 96 (8 x 12) wells
  • Lyophilized Mouse IFNγ standards, 10ng x 2
  • Sample/Standard diluent buffer, 30mL
  • Biotin conjugated anti-Mouse IFNγ antibody (Concentrated), Dilution: 1:100, 130μL
  • Antibody diluent buffer, 12mL
  • Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated), Dilution: 1:100, 130μL
  • ABC diluent buffer, 12mL
  • TMB substrate, 10mL
  • Stop solution, 10mL
  • Wash buffer (25X), 30mL

Use to measure quantitatively mouse Ifng.

This kit was based on sandwich enzyme-linked immune-sorbent assay technology

Specifications

Calibration Range 31.2 to 2000pg/mL
Conjugate Biotin
Cross Reactivity Mouse
Description Ifng (Mouse) ELISA Kit is intended for the quantitative measurement of mouse Ifng.
Detection Limit 5pg/mL
Detection Method Colorimetric
Includes One 96-well plate pre-coated with anti-Mouse IFNΓ antibody (96 (8 x 12) wells), Lyophilized Mouse IFNΓ standards (10ng x 2), Sample/standard diluent buffer (30mL), Biotin conjugated anti-Mouse IFNΓ antibody (Concentrated), Dilution: 1:100 (130μL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated), Dilution: 1:100 (130μL), ABC diluent buffer (12mL), TMB substrate (10mL), Stop solution (10mL), 30mL wash buffer (25X)
Regulatory Status For research use only (RUO)
Sample Type Body Fluid, Cell Culture Supernate, Serum, Tissue Lysate
Sample Volume 100 μL
Sensitivity <5 pg/mL
Packaging 96-well
Content And Storage Store at 4°C for 4 months, or at -20°C for 8 months.
For Use With (Application) Quantification
Product Type Ifng (Mouse) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.