Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ IgA (Cat) ELISA Kit

Catalog No. 89139341 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-139-341 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-139-341 Supplier Abnova™ Supplier No. KA4513
Add to Cart
Add to Cart

Sandwich enzyme immunoassay for the quantitative measurement of cat IgA in plasma and serum samples

  • Sample Type: Plasma, serum
  • Sample Volume: 100μL
  • Conjugate: HRP
  • Reactivity: Cat
  • Detection Method: Colorimetric
  • Calibration Range: 12.5 to 400 ng/mL
  • Applications: Quantification
  • Storage: Store the kit at 4°C.

Specifications

Calibration Range 12.5 to 400 ng/mL
Description IgA (Cat) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of cat IgA.
Detectable Analytes IgA
Assay Sandwich ELISA
Conjugate Biotin
Detection Method Colorimetric
Product Type ELISA Kit
Sample Type Plasma, Serum
Sufficient For 96 Assays
For Use With (Application) Quantification
Host Species Cat
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Volume 100 μL
Storage Requirements Store the kit at 4°C.
Show More Show Less

For research use only (RUO).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.