Learn More
Description
Includes
- One 96-well plate pre-coated with anti-Human IL-17 antibody 96 (8x12) wells
- Lyophilized IL-17 standards 10ng x 2
- Sample / Standard diluent buffer 30mL
- Biotin conjugated anti-Human IL-17 antibody (Concentrated), Dilution: 1:100 130 ul
- antibody diluent buffer 12mL
- Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated), Dilution: 1:100 130 ul
- ABC diluent buffer 12mL
- TMB substrate 10mL
- Stop solution 10mL
- Wash buffer (25X) 30mL
- Kit is based on sandwich enzyme-linked immune-sorbent assay technology
- Anti-IL-17 polyclonal antibody was pre-coated onto 96-well plates
- Read the O.D. absorbance at 450 nm in a microplate reader, and then the concentration of IL-17 can be calculated
- Store at 2-8°C for 4 months, or at -20°C for 8 months
- For research use only
Specifications
Specifications
| Calibration Range | 15.6 to 1000pg/mL |
| Description | IL17A (Human) ELISA Kit is intended for the quantitative measurement of human IL17A. |
| Conjugate | Biotin |
| Product Type | IL17A (Human) ELISA Kit |
| Sample Type | Body Fluids, Cell Culture Supernatant, Plasma, Serum, Tissue Lysates |
| Target Species | Human |
| For Use With (Application) | Quantification |
| Gene ID (Entrez) | 3605 |
| Gene Symbol | IL17A |
| Quantity | 1 Kit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.