Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ IL1RL1 (Human) ELISA Kit

Catalog No. 89100421 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-421 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-421 Supplier Abnova™ Supplier No. KA2141

Includes

  • IL-1 R4 Microplate: 96 wells coated with anti-Human IL-1 R4
  • Wash Buffer Concentrate (20x)
  • Standard Protein: Human IL-1 R4. 1 vial is enough to run each standard in duplicate
  • Detection Antibody IL-1 R4: Biotinylated anti-Human IL-1 R4. Each vial is enough to assay half the microplate
  • HRP-Streptavidin concentrate: 300x concentrated HRP-conjugated Streptavidin
  • TMB One-Step Substrate Reagent: 3, 3', 5, 5'- tetramethylbenzidine (TMB) in buffered solution
  • Stop Solution: 0.2M sulfuric acid
  • Assay Diluent A: Diluent buffer, containing 0.09% sodium azide as preservative
  • Assay Diluent B: 5x concentrated buffer, 15mL

Use to measure quantitatively human IL1RL1. Enzyme-linked immunosorbent assay.

This assay employs an antibody specific for human IL-1 R4 coated on a 96-well plate

Specifications

Calibration Range 1.65 to 1200pg/mL
Conjugate HRP
Cross Reactivity Human
Description IL1RL1 (Human) ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative measurement of human IL1RL1.
Detection Limit 2pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Serum, Plasma, Cell Culture Supernatant
Sample Volume 100 μL
Sensitivity 2 pg/mL
Packaging 96-well
Content And Storage Store the kit at -20°C. For best results, please store the components as described in the protocol. Avoid repeated freezing and thawing.
For Use With (Application) Quantification
Product Type IL1RL1 (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.