Learn More
Description
Includes
- Lyophilized recombinant Human IL-1RA standard, 10ng/tube x 2
- One 96-well plate precoated with anti-Human IL-1RA antibody, 1 in no.
- Sample diluent buffer, 30mL
- Biotinylated anti- Human IL-1RA antibody, dilution 1:100, 130μLl
- Antibody diluent buffer, 12mL
- Avidin-Biotin-Peroxidase Complex (ABC), dilution 1:100, 130μL
- ABC diluent buffer, 12mL
- TMB color developing agent, 10mL
- TMB stop solution, 10mL
Specifications
Specifications
| Calibration Range | 31.2 to 2000pg/mL |
| Conjugate | Biotin |
| Cross Reactivity | Human |
| Description | Human IL1RN ELISA kit is a sandwich enzyme immunoassay for the quantitative measurement of human IL1RN. |
| Detection Limit | 2pg/mL |
| Detection Method | Colorimetric |
| Includes | 96-well Antibody coated microwell, 2 vials x 10ng recombinant Human IL1RN standard (lyophilized), 30mL Sample Diluent Buffer, 130μL Biotinylated Anti-Human IL-1RA Antibody, 12mL Antibody Diluent Buffer,, 130μL Avidin-Bi |
| Quantity | 1 Kit |
| Sample Type | Body Fluid, CCS, Serum, Tissue Lysates |
| Sample Volume | 100 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.