Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ IL7 (Human) ELISA Kit

Catalog No. 89100203 Shop All Abnova Corporation Products
Change view
Click to view available options
Calibration Range:
1.5 to 50pg/mL
15.6 to 1000pg/mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Calibration Range
89-100-203 1.5 to 50pg/mL
89-028-198 15.6 to 1000pg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-203 Supplier Abnova™ Supplier No. KA0525
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of IL-7

Specifications

Calibration Range 1.5 to 50pg/mL
Conjugate HRP
Cross Reactivity Human
Description IL7 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human IL7.
Detection Limit 1pg/mL
Detection Method Colorimetric
Includes 96 wells-antibody coated microwells, Biotin conjugated (7mL), Avidin conjugate (12mL), IL-7 Standard 4ng/vial x 2 vial, Calibrator Diluent I (25mL), Calibrator Diluent II (25mL), Wash Buffer 20X 60mL, Substrate A (11mL), Substrate B (11mL), Stop Solution 14mL
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type CCS, Plasma, Serum
Sample Volume 100 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type IL7 (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.