Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Insulin (Human) ELISA Kit

Catalog No. 89024462 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
96 Assays
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-462 96 Assays
89-101-131 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-024-462 Supplier Abnova™ Supplier No. KA0921

Includes

  • Microwells coated with Insulin MAb 12x8x1
  • Insulin Standard 1:1 vial (Ready to use) 2mL
  • Insulin Standards 2-6: 5 vials (Ready to use) 1mL
  • Insulin Enzyme Conjugate: 1 vial 1mL
  • Assay Diluent: 1 bottle (Ready to use) 14mL
  • TMB Substrate: 1 bottle (Ready to use) 12mL
  • Stop Solution: 1 bottle (Ready to use) 12mL
  • 20X Wash concentrate: 1 bottle 25mL

For quantitative measurement Insulin in Human Serum or Plasma

Specifications

Calibration Range 6.25 to 100 μIU/mL
Conjugate HRP
Cross Reactivity Human
Detection Limit 1.467μIU/mL
Detection Method Colorimetric
Includes 96-well microwell coated with Insulin MAb, 2mL Insulin standard 1, 1mL Insulin standards 2, 4 x 0.5mL Insulin standards 3-6, 0.5mL Insulin Enzyme Conjugate, 14mL Assay diluent, 12mL TMB Substrate, 25mL 20X wash concentrate, 12mL TMB solution,, 12mL stop solution
Quantity 96 Assays
Sample Type Plasma, Serum
Sample Volume 25 μL
Content And Storage 4°C
For Use With (Application) Quantitative measurement Insulin in Human Serum or Plasma, quantification
Product Type Insulin (Human) ELISA kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.