Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Iron Assay Kit

Catalog No. 89028379 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-379 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-379 Supplier Abnova™ Supplier No. KA1647
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Reagent A, 50mL
  • Reagent B, 4mL
  • Reagent C, 4mL
  • Iron standard: 10mg/dL Fe2+, 1mL

For quantitative colorimetric iron determination at 590nm in biological sample, serum

Specifications

Calibration Range 4.8 to 179μM
Content And Storage Store reagent A at room temperature and other reagents at 4°C.
Description Iron Assay Kit is a quantitative colorimetric iron determination at 590 nm.
Detection Method Colorimetric
For Use With (Application) Functional Study
Includes 50mL Reagent A, 4mL reagent B, 4mL reagent C, 1mL Iron standard, 10mg/dL Fe2+
Quantity 1 Kit
Sample Type Biological Sample, Serum
Sample Volume 50 μL
Type Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.