Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ JARID Demethylase Activity/Inhibition Assay Kit (Fluorometric)

Catalog No. 89028223 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-223 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-223 Supplier Abnova™ Supplier No. KA1529
Add to Cart
Add to Cart

Includes

  • JD1 (10x Wash Buffer), 14mL
  • JD2 (JARID Assay Buffer), 4mL
  • JD3 (JARID Substrate, 50μg/mL), 60μL
  • JD4 (JARID Assay Standard, 50μg/mL), 10μL
  • JD5 (Capture Antibody, 1000μg/mL) 5μL
  • JD6 (Detection Antibody, 400μg/ml), 6μL
  • JD7 (Fluoro-Developer), 10μL
  • JD8 (Fluoro-Enhancer), 10μL
  • JD9 (Fluoro-Dilution), 4mL
  • Co-factor 1, 30μL
  • Co-factor 2, 30μL
  • Co-factor 3, 30μL
  • Six 8-well wssay strips (with frame)
  • Adhesive covering film

To screen JARID demethylase inhibitors, functional study

Specifications

Content And Storage Store JD1, JD5, JD8, Co-factor 1, Co-factor 2, Co-factor 3 and 8-Well Assay Strips at 4°C away from light. Store JD3, JD4, JD6, and JD7 at -20°C away from light.Store remaining components at room temperature.
Description JARID Demethylase Activity/Inhibition Assay Kit (Fluorometric) is use for screening JARID demethylase inhibitors.
Detection Method Fluorometric
For Use With (Application) Functional Study
Includes 14mL JD1 (wash buffer 10X), 4mL JD2 (JARID assay buffer), 60μL JD3 (JARID substrate, 50μg/mL), 10μL JD4 (JARID assay standard, 50μg/mL), 5μL JD5 (Capture antibody, 1
Quantity 1 Kit
Type Activity/inhibition Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.