Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ JC-1 Mitochondrial Membrane Potential Assay Kit

Catalog No. 89024610 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-610 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-610 Supplier Abnova™ Supplier No. KA1324
Add to Cart
Add to Cart

Fluorescence-based method for tracing the behavior of mitochondria

Immunofluorescence, Functional Study, Flow Cytometry

Specifications

Content And Storage Store the kit at -20°C temporarily. For best results, store the components as described in the protocol.
Description JC-1 Mitochondrial Membrane Potential Assay Kit is a fluorescence-based method for tracing the behavior of mitochondria.
Detection Method Fluorometric
For Use With (Application) Flow Cytometry, Functional Study
Includes 500μL JC-1 reagent, 3 tablets buffer tablet
Quantity 1 Kit
Type Membrane Potential Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.