Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Ketone Body Assay Kit

Catalog No. 89028362 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-362 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-362 Supplier Abnova™ Supplier No. KA1630
Add to Cart
Add to Cart

Includes

  • Acetoacetic acid buffer, 20mL
  • Acetoacetic acid reagent, 1mL
  • Acetoacetic acid standard (80mM), 200μL
  • HBDH enzyme, 120μL
  • 3-hydroxybutyric acid buffer, 20mL
  • 3-hydroxybutyric acid reagent, 1mL
  • 3-hydroxybutyric acid standard (80mM), 200μL

Ketone Body Assay Kit is a quantitative colorimetric determination of ketone body at 340 nm.

  • Sensitive and accurate: Uses 10 μL sample.
  • Linear detection range of 0.12 to 8 mM for each ketone body in 96-well plate assay
  • Convenient: The procedure involves adding a single working reagent, and reading the optical density at room temperature
  • High-throughput: Can be automated as a high-throughput 96-well plate assay for many samples per day
  • Store all reagents at -20°C; shelf life of 6 months after receipt
  • For research use only

Specifications

Description Ketone Body Assay Kit is a quantitative colorimetric determination of ketone body at 340 nm.
For Use With (Application) Functional Study
Includes 20mL Aceto acetic Acid Buffer, 1mL Aceto acetic Acid reagent, 200μL Aceto acetic Acid Standard (80mM), 120μL HBDH Enzyme, 20mL 3-Hydroxybutyric Acid Buffer, 1mL 3-Hydroxybutyric Acid reagent
Sample Volume 10μL
Content And Storage Store the kit at at -20°C.
Product Type Keto ne body assay kit
Quantity Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.