Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Kinase Assay Kit

Catalog No. 89028363 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-363 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-363 Supplier Abnova™ Supplier No. KA1631
Add to Cart
Add to Cart

Generic fluorometer high-throughput kinase assay in biological sample, cell

Specifications

Content And Storage Store the kit at -20°C.
Description Kinase Assay Kit is a generic fluorimetric high-throughput kinase assay.
Detection Method Fluorometric
For Use With (Application) Functional Study
Includes 10mL Reagent A, 10mL reagent B, 25mL assay buffer,100μL standard (3mM ADP)
Quantity 1 Kit
Sample Type Biological Sample, Cell
Type Kinase Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.