Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ L-Alanine Assay Kit

Catalog No. 89101190 Shop All Abnova Corporation Products
Change view
Click to view available options
Calibration Range:
Colorimetric: 1 to 200μM, Fluorimetric: 0.4 to 20μM
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Calibration Range
89-101-190 Colorimetric: 1 to 200μM, Fluorimetric: 0.4 to 20μM
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-101-190 Supplier Abnova™ Supplier No. KA3773
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Alanine assay buffer, 25mL
  • Alanine probe (in DMSO), 0.2mL
  • Alanine converting enzyme (lyophilized), 1 vial
  • Alanine development mix (lyophilized), 1 vial
  • Alanine standard (10μmol, lyophilized), 1 vial

Used for detecting alanine and functional study

Specifications

Calibration Range Colorimetric: 1 to 200μM, Fluorimetric: 0.4 to 20μM
Content And Storage Store the kit at -20°C.
Description L-Alanine Assay Kit is a quantitative colorimetric/fluorimetric determination of L-alanine at OD570, FL530/585 nm.
Detection Method Colorimetric, Fluorometric
For Use With (Application) Functional Study
Includes Developer (6mL), ALT Enzyme (120μL), Dye Reagent (120μL), Cosubstrate (600μL), Alanine Standard (400μL )
Quantity 1 Kit
Sample Type Culture Medium, Plasma, Serum, Tissue, Urine
Sample Volume 50 μL
Type Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.