Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ L-Lactate Assay Kit (Fluorometric)

Catalog No. 89101192 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-101-192 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-101-192 Supplier Abnova™ Supplier No. KA3776
Add to Cart
Add to Cart

Requires

  • Multi-channel pipetting devices
  • Black, flat-bottom 96-well plates
  • Fluorescent plate reader capable of reading at λex/em≥530/585nm

One kit sufficient for 100 assays. L-Lactate Assay Kit is a quantitative fluorimetric determination of L-lactate at FL530/585nm.

  • Convenient: Room temperature assay. Read fluorescence 60 minutes after adding working reagent.
  • High-throughput: Easily automated as a high-throughput 96-well plate assay for thousands of sample per day.

Specifications

Calibration Range 1 to 50μM
Content And Storage Store the kit at -20°C.
Description L-Lactate Assay Kit is a quantitative fluorimetric determination of L-lactate at FL530/585 nm.
Detection Method Fluorometric
For Use With (Application) Functional Study
Includes Assay Buffer, NAD Solution, Probe, Enzyme A, Enzyme B, Standard
Quantity 1 Kit
Sample Type Culture Medium, Plasma, Serum
Sample Volume 50 μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.