Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Lactose Assay Kit

Catalog No. 89028404 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-404 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-404 Supplier Abnova™ Supplier No. KA1672
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For quantitative colorimetric lactose determination in biological sample, milk

Specifications

Calibration Range 17 to 2000μM colorimetric assay, 6 to 100μM fluorimetric assay
Content And Storage Store the kit at -20°C.
Description Lactose Assay Kit is a quantitative colorimetric lactose determination.
Detection Method Colorimetric
For Use With (Application) Functional Study
Includes 10mL assay buffer, enzyme mix dried, Lactase dried, 120μL dye reagent, 1mL standard (20mM Lactose)
Quantity 1 Kit
Sample Type Biological Sample, Milk
Sample Volume 20 μL
Type Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.