Learn More
Specifications
Specifications
| Calibration Range | 0.3 to 10ng/mL |
| Conjugate | HRP |
| Cross Reactivity | Human |
| Description | LCN2 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human lipocalin-2. |
| Detection Limit | 0.02ng/mL |
| Detection Method | Colorimetric |
| Includes | 96 wells- antibody coated microwell strips, Biotin labelled antibody conc.20X (0.70mL), Streptavidin-HRP Conjugate (13mL), Master standard (2 vials), Quality control HIGH (2 vials), Quality control LOW (2 vials), Diluent buffer (20mL), Biotin-Ab diluent (13mL), Wash solution (100mL), Substrate Reagent (13mL), Stop Solution (13mL) |
| Quantity | 1 Kit |
| Regulatory Status | For research use only (RUO) |
| Sample Type | Plasma (Citrate, EDTA, Heparin), Serum, Urine |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.