Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ LCN2 (Human) ELISA Kit

Catalog No. 89100713 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CCS, Plasma, Serum, Tissue Sample, Urine
Culture Supernatant
Plasma (Citrate, EDTA, Heparin), Serum, Urine
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-713 Culture Supernatant
89-100-796 CCS, Plasma, Serum, Tissue Sample, Urine
89-100-014 Plasma (Citrate, EDTA, Heparin), Serum, Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 89-100-713 Supplier Abnova™ Supplier No. KA2530

Sandwich Enzyme Immunoassay for the quantitative measurement of Human NGAL/Lipocalin-2 in serum, plasma, urine, tissue culture medium, and other biological media.

Specifications

Calibration Range Not Applicable
Conjugate HRP
Cross Reactivity Human
Description LCN2 (Human) ELISA Kit is used for the quantitative measurement of human LCN2.
Detection Limit Not Applicable
Detection Method Colorimetric
Includes NGAL standard (200uL), Human NGAL recombinant, (200uL), Anti -NGAL capture antibody (60uL), Anti-NGAL detection antibody (110uL),
Streptavidin conj. peroxidase (110uL), Microwell strips, Coating Buffer (12mL), Blocking Agent (24mL), Sample Diluent (50mL), Wash Concentrate (50mL), SA-HRP Diluent (15mL), Substrate Solution (20mL), Stop solution (20mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Culture Supernatant
Sample Volume 100 μL
Content And Storage Store standard, capture antibody, detection antibody and Streptavidin conj. Peroxidase at -20°C; others at 4°C.
For Use With (Application) Quantification
Product Type LCN2 (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.