Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ LDL Uptake Cell-Based Assay Kit

Catalog No. 89024611 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-611 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-611 Supplier Abnova™ Supplier No. KA1327
Add to Cart
Add to Cart

Includes

  • Cell-based assay fixative, 1 vial
  • Rabbit anti-LDL receptor primary, 1 vial
  • Cell-based assay blocking solution, 1 vial
  • LDL-DyLightTM 549, 1 vial
  • DyLightTM 488-conjugated goat anti- rabbit IgG secondary antibody, 1 vial

For studying LDL uptake and regulation at the cellular level, immunofluorescence, functional study

Specifications

Content And Storage Store the kit at 4°C temporarily. For best results, store the components as described in the protocol.
Description LDL Uptake Cell-Based Assay Kit is a tool for studying LDL uptake and regulation at the cellular level.
For Use With (Application) Functional Study
Includes 1 vial fixative, 1 vial rabbit anti-LDL receptor primary, 1 vial blocking solution, 1 vial LDL-DyLight 549, 1 vial DyLight 488 conjugated goat anti-rabbit IgG secondary antibody
Quantity 1 Kit
Type LDL Uptake Cell Based Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.