Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Lipase Assay Kit

Catalog No. 89028386 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-386 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-386 Supplier Abnova™ Supplier No. KA1654
Add to Cart
Add to Cart

Colorimetric determination of lipase activity at 412nm in biological sample, plasma, saliva, serum, urine

Specifications

Calibration Range 40 to 1600U/L
Content And Storage Store the kit at 4°C.
Description Lipase Assay Kit is a rapid colorimetric determination of lipase activity at 412 nm.
Detection Method Colorimetric
For Use With (Application) Functional Study
Includes 15mL assay buffer (pH 8.5), 530mg color reagent, 1mL BALB reagent, 2mL (equivalent to 735U/L) calibrator
Quantity 1 Kit
Sample Type Biological Sample, Plasma, Saliva, Serum, Urine
Sample Volume 10 μL
Type Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.