Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Lipid Hydroperoxide (LPO) Assay Kit

Catalog No. 89024612 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-612 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-612 Supplier Abnova™ Supplier No. KA1328
Add to Cart
Add to Cart

Used for detecting hydroperoxides and functional study

Specifications

Content And Storage Store the kit at -80°C temporarily. For best results, store the components as described in the protocol.
Description Lipid Hydroperoxide (LPO) Assay Kit is a measurement of detecting hydroperoxides.
For Use With (Application) Functional Study
Includes 1 vial LPO assay FTS reagent 1, 1 vial LPO assay FTS reagent 2, 1 vial Lipid Hydroperoxide standard, 1 vial LPO assay extract R, 1 vial LPO assay Triphenylphosphine, 1 plate 96-well solid plate (Glass), 1 cover foil plate cover
Quantity 1 Kit
Type Detection Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.